Would you like to log in again?
Pierce™ Kinase Enrichment Kit, Thermo Scientific
Supplier: Thermo Scientific
Pierce
88310 88312 88312 88310
PI88312EA 1108.8 USD InStock
CAPI88310 CAPI88312 PI88312 PI88310
Pierce™ Kinase Enrichment Kit, Thermo Scientific
Assays Enzyme Assays
The Pierce™ Kinase Enrichment Kits utilise an ActivX™ ATP or ADP probe to covalently label the active site of ATPases, including chaperones and metabolic enzymes, to enable their selective enrichment using a desthiobiotin tag.
  • Labels Only the Conserved Active-Site Lysines of Nucleotide-Binding Proteins
  • Flexible use for in vitro Labeling of ATPase Enzymes Derived from Cells or Tissues
  • Compatible with use with Western Blot or Mass Spectrometry (MS) Workflows
ActivX™ probes feature an amine-reactive nucleotide analog and a desthiobiotin (biotin analogue) tag that facilitates selective labeling of lysines in the kinase active site and then subsequent enrichment and recovery of labelled protein.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty