Would you like to log in again?
Pierce™ Triple Quadrupole Calibration Solution
Supplier: Thermo Scientific
Pierce™
88325 88325
PI88325EA 357.97 USD InStock
CAPI88325 PI88325
Pierce™ Triple Quadrupole Calibration Solution
Standards Color Reference and Spectroscopy Standards
This triple Quadrupole Calibration Solution is a mixture of high purity, ionizable components specifically designed for positive mode calibration of triple stage quadrupole instruments.
  • High-Purity - Mass Spectrometry Grade Reagents in a Non-Leachable Container
  • Strong Peaks - Mixture of Three Tyrosine Polymers in Methanol/Formic Acid Solution
  • Stable - Store at 2 to 8 °C for More than One Year
Pierce Triple Quadrupole Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the required routine calibration to maintain the robust performance of Thermo Scientific Mass Spectrometers
Pierce Triple Quadrupole Calibration Solution can be used to calibrate the TSQ Quantum series, TSQ Discovery series, TSQ Quantum Access Series and TSQ Vantage Series Mass Spectrometer instruments.
The Triple Quadrupole Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty