Would you like to log in again?
96-Well Plates for Pierce™ Quantitative Fluorometric Peptide Assay, Thermo Scientific
Supplier: Thermo Scientific
88378
PI88378PK 211.69 USD InStock
PI88378
96-Well Plates for Pierce™ Quantitative Fluorometric Peptide Assay, Thermo Scientific
Assays Protein Assays
Thermo Scientific™ 96-Well Plates are manufactured using high-quality polystyrene and are high-binding with black, flat well bottoms to accommodate fluorescence assays such as the Pierce™ Quantitative Fluorometric Peptide Assay.
Ordering information: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty