Would you like to log in again?
Pierce™ Cell Culture Medium for SILAC, Thermo Scientific
Supplier: Thermo Scientific
Pierce
88425 88425 88441 88426 88441 88424 88424 88426
PI88441EA 97.14 USD InStock
CAPI88441 CAPI88424 CAPI88426 CAPI88425 PI88441 PI88426 PI88425 PI88424
Pierce™ Cell Culture Medium for SILAC, Thermo Scientific
Cell Culture Media
The Pierce™ cell culture media for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).
  • Sterile, Endotoxin-Free and Cell Culture-Compatible
  • Flexible with Liquid Media Deficient in Both L-Lysine and L-Arginine
Allows more complete proteome coverage through multiple isotopic amino acid labeling. This is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty