Would you like to log in again?
Pierce™ Streptavidin Magnetic Beads, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pierce
88817 88817 88816 88816
PI88817EA 1468.55 USD InStock
PI88817 PI88816 CAPI88817 CAPI88816
Pierce™ Streptavidin Magnetic Beads, Thermo Scientific
Beads Magnetic Beads
Thermo Scientific Pierce Streptavidin Magnetic Beads accelerate throughput for automated magnetic purification of biotinylated molecules.
  • High-Performance Beads—non-aggregating, Pre-Blocked, Iron Oxide, Superparamagnetic Microparticles Provide Exceptional Uniformity for Automated HTS and Manual Applications Alike
  • Stable Immobilization Chemistry—Streptavidin is Immobilized using Leach-Resistant Chemistry
  • High Capacity—Superior Quality Beads with High Binding Capacity Provide Rapid and Efficient Biomolecule Purification from Complex Samples
  • Low Non-Specific Binding—Stable, Pre-Blocked Beads Provide Clean Purification Products (e.g., Antigen Eluted in IP with Biotinylated Antibody) that are Compatible with Mass Spectrometry Analysis and other Downstream Analyses
  • Superior Performance—Nearly Three Times Higher Binding Capacity than Typical Beads from other Suppliers, Allowing the use of Smaller Amounts Per Experiment
These streptavidin magnetic beads are validated and optimized for use with high-throughput magnetic platforms, such as the Thermo Scientific KingFisher 96 and KingFisher Flex Instruments, but the beads also enable premium performance for simple benchtop applications using an appropriate magnetic stand
Pierce Streptavidin Magnetic Beads use a recombinant form of streptavidin with a mass of 53kDa and a near-neutral isoelectric point (pI). The protein is a tetramer having four biotin-binding sites. Unlike avidin, streptavidin has no carbohydrate groups, resulting in low nonspecific binding. The high-affinity interaction between streptavidin and biotin cannot be dissociated efficiently except with very harsh conditions, such as boiling in sample loading buffer for SDS-PAGE or 8M guanidine∙HCl, pH 1.5. Consequently, it is often possible to elute binding partners in an interaction complex without also eluting the biotinylated component.
The iron oxide, super-paramagnetic particles offer superior performance (high capacity and low nonspecific binding) compared with other commercial magnetic beads.
Ordering information: Upon receipt store at 4?C. Product is shipped on an ice pack.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty