Would you like to log in again?
Pierce™ Direct Magnetic IP/Co-IP Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce™
88828 88828
PI88828EA 680.49 USD InStock
PI88828 CAPI88828
Pierce™ Direct Magnetic IP/Co-IP Kit, Thermo Scientific
Protein Purification Protein Purification Kits
The direct magnetic IP/Co-IP kit uses NHS-chemistry to covalently immobilize IP antibodies onto magnetic beads for effective immunoprecipitation and co-immunoprecipitation.
  • Antibody-free IP-antibody is irreversibly attached to the magnetic beads, resulting in negligible co-elution of intact antibody or heavy and light chains with the purified antigen
  • Fast-complete antibody conjugation and immunoprecipitation in less than 4 hours
  • Convenient-the IP/co-IP kit contains magnetic beads and all essential buffers for optimal immunoprecipitation
  • Antibody compatible-use with any antibody species, class or isotype
  • Low nonspecific binding-the bead surface is pre-blocked and all nonreacted NHS-ester groups are fully quenched
  • Protocol-compatible-protein coupling to the beads and immunoprecipitation can be performed manually or by automation (e.g., thermo scientific kingFisher instruments)
  • Scalable-use only the amount of antibody needed for a single IP experiment or prepare a larger quantity of antibody magnetic beads for multiple experiments
Magnetic IP with NHS chemistry is convenient and allows for the immobilization of IP antibody from any species and subtype without the variations in coupling efficiency seen with Protein A and Protein G. The IP antibody is covalently attached so target proteins or co-IP protein complexes can be eluted and analyzed without antibody contamination. The kit contains an optimized protocol and all buffers and reagents necessary for antibody coupling and high yield IP or co-IP using either manual magnetic stands or automated platforms.
The activated magnetic beads contain N-hydroxysuccinimide (NHS) functional groups, which react with primary amines on antibodies to form stable amide linkages. The antibody is first coupled to the NHS-activated magnetic beads in an amine-free buffer. The antibody-coupled beads are washed to remove any non-covalently bound antibody and quenched. The prepared beads are incubated with the antigen sample, washed to remove non-bound material and eluted in a low-pH elution buffer to dissociate the antigen. The kit includes optimised buffers that minimise non specific binding and maximise antigen yield. Lane marker sample buffer is included for preparing samples for SDS-PAGE analysis. The beads are manually removed from the solution with a magnetic stand or by automation.
Delivery information: The kit includes NHS-activated magnetic beads, 1 ml; IP lysis/wash buffer, 2×50 ml; elution buffer (pH 2.0), 5 ml; lane marker sample buffer, non-reducing, (5×), 5 ml; neutralisation buffer (pH 8.5), 0.5 ml; 0.67 M borate buffer, 1 ml, BupH borate buffer pack, 1 pack; quenching buffer, 25 ml.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty