Would you like to log in again?
Pierce™ Anti-HA Magnetic Beads
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Pierce™
88836 88836 88837 88837
PI88837EA 1381.26 USD InStock
PI88837 PI88836 CAPI88837 CAPI88836
Pierce™ Anti-HA Magnetic Beads
Beads Magnetic Beads
Pierce™ Anti-HA magnetic beads are affinity particles for immunoprecipitation of recombinant HA-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.
  • Highly specific anti-HA monoclonal antibody (clone 2-2.2.14) enables high yield and high purity immunoprecipitation
  • Product instructions provide an easy-to-follow, optimized protocol for immunoprecipitation in approximately one hour
  • Beads are compatible with manual and automated workflows
Stable, pre-blocked beads and specific antibody minimize off-target binding for HA-tag IP or co-IP experiments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty