Would you like to log in again?
Pierce™ 1-Step Human In Vitro Protein Expression Kits, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce Pierce™
88881 88882 88882 88881
PI88882EA 1087.48 USD InStock
CAPI88882 CAPI88881 PI88882 PI88881
Pierce™ 1-Step Human In Vitro Protein Expression Kits, Thermo Scientific
Protein Expression Protein Expression Systems
Thermo Scientific 1-Step Human In Vitro Protein Expression Kits enable the translation and post-transcriptional modification of full-length proteins from mRNA or plasmid templates with yields of up to 100 µg/ml per reaction. Express full-length functional proteins in 90 minutes using the coupled human cell-free protein expression system.
  • Functional - uses the human translational machinery to express active proteins
  • Convenient - perform transcription and translation in a single step
  • High performance - greater yields compared to rabbit reticulocyte in vitro translation
  • Reliable - express proteins that fail in rabbit reticulocyte systems
The Human IVT Kits are unique HeLa cell lysate-based protein expression systems for in vitro translation or coupled transcription/translation reactions.
Requires 30 °C incubator or water bath.
The 1-Step Coupled Human IVT Kit for DNA is a cell-free system using the cellular transcription and translation machinery from a modified HeLa cell line. The procedure is quick and easy and is an effective alternative to other protein expression methods. Simply add an appropriate expression construct to a mixture of HeLa cell lysate, Accessory Protiens, Reaction Mix and incubate at 30 °C for 90 minutes to overnight.
The 1-Step Human IVT Kit for mRNA also uses the human protein translation machinery to produce functional protein. This kit is recommended for translation of mRNA transcripts containing an ECMV IRES element and mRNA stabilizing features designed into the pT7CFE1 expression vector.
Delivery information: Each kit contains HeLa cell lysate (110 or 500 µl), Accessory proteins (25 or 100 µl), Reaction mix (40 or 200 µl), Nuclease-free water (1.5 ml), Expression vector (pT7CFE1-CHis - 10 µg) and a GFP-positive control vector (pCFE-GFP - 10 µg).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty