Would you like to log in again?
Pierce™ Azide Labeled Sugars, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
88904 88905 88903 88903 88905 88904
PI88903EA 504 USD InStock
PI88903 PI88905 PI88904 CAPI88903 CAPI88905 CAPI88904
Pierce™ Azide Labeled Sugars, Thermo Scientific
Protein Linking and Labeling Systems
Thermo Scientific Pierce™ Azide-Labeled sugars provide a highly specific approach for studying glycoproteins through in vivo metabolic labeling and chemoselective ligation.
  • Bioorthogonal - The Azido-Sugar Compounds do not Interfere with Endogenous Cellular Pathways and Substitute for their Naturally Occurring Analogues
  • Reaction Chemistry with Phosphine Compounds Occurs Effectively in Simple Buffer Conditions; Requires no Accessory Reagents Such as Copper or Reducing Agents
  • Azide and Phosphine Groups do not React or Interfere with Components of Biological Samples but Conjugate to One Another with High Efficiency
  • Azide Tag can be Targeted for Detection, Immobilization, Conjugation or Affinity Purification Depending on Which Phosphine-Activated Compound it is Reacted with
These sugars are Azide-Derivatives of naturally occurring monosaccharides that cells use to glycosylate proteins using post-translational modification biochemical pathways. When supplied to cells, these compounds become incorporated by glycosylation events to effectively "tag" glycoproteins with the azide group. The azide group then can be specifically targeted for detection or conjugation using alkyne-activated reagents. When used in combination with phosphine-activated fluorescent dyes, biotin reagents, and or other compounds, these azido-modified sugars facilitate the investigation of cellular pathways involving glycosylation.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty