Would you like to log in again?
Pierce™ GlycoLink™ Immobilization Ki, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce™
88941 88941
PI88941EA 1041.6 USD InStock
CAPI88941 PI88941
Pierce™ GlycoLink™ Immobilization Ki, Thermo Scientific
Protein Purification Protein Purification Kits
The GlycoLink™ Immobilisation Kit contains affinity columns and reagents to immobilise glycoproteins through oxidised sugar groups for use in affinity-purification applications or studies of protein-protein interactions. This kit is ideal for purification of antigens using covalently coupled antibodies, determining binding partners for a glycoprotein of interest and for performing large-scale affinity purifications.
  • High capacity with 1 to 10 mg of oxidised antibody or other glycoprotein per ml of resin
  • Immobilise in as little as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours
  • Hydrazide-activated UltraLink resin conjugates only to purified glycoproteins whose sugar (eg. sialic acid) groups have been gently oxidised with periodate
  • Resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilisation step required
  • Immobilises IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target
Antibodies immobilised by this hydrazide immobilisation chemistry have unobstructed antigen-binding sites and optimal purification capability. These columns can be regenerated and reused five times without loss in binding capacity.
Delivery information: Kit contains GlycoLink™ columns (1 ml, 10 columns), GlycoLink™ Coupling Buffer (2×100 ml), aniline (0.2 ml), Wash Buffer (120 ml), sodium meta-periodate (0.5 g), Zeba™ spin desalting columns (5 ml, 10 columns), PBS pack (makes 500 ml, 1 pack) and column accessories (10 each).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty