Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GlycoLink™, Pierce™ Immobilization Kits and Resin, Thermo Scientific

Supplier: Thermo Fisher Scientific
GlycoLink™
88942 88942
PI88942EA 620.31 USD InStock
CAPI88942 PI88942
GlycoLink™, Pierce™ Immobilization Kits and Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Hydrazide-activated resin to immobilize glycoproteins for affinity purification.

  • High-capacity—immobilize 1 to 10mg of oxidized antibody or other glycoprotein per milliliter of resin
  • Fast and efficient—immobilize in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)
  • Specific immobilization—the hydrazide-activated UltraLink Resin conjugates only to purified glycoproteins whose sugar (e.g., sialic acid) groups have been gently oxidized with periodate
  • Stable attachment—resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilization step is required
  • Preserves antibody function—immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target
  • Convenient sizes—choose from two complete 10-column kits: one with 1mL columns and one with 0.1mL columns

Thermo Scientific GlycoLink Immobilization Kits provide affinity columns and reagents to immobilize glycoproteins through oxidized sugar groups for use in affinity-purification applications or studies of protein-protein interactions.

The GlycoLink Kits use hydrazide-activated resin and a special catalyst to prepare affinity columns with polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Any glycosylated proteins (even monoclonal antibodies) that contain periodate-oxidizable carbohydrate groups can be immobilized with these kits to make reusable affinity columns. Antibodies immobilized by this hydrazide immobilization chemistry have unobstructed antigen-binding sites and optimal purification capability. Prepared columns with stable glycoproteins or antibodies can be regenerated and reused at least five times for affinity-purification without significant loss in binding capacity.

Kit Content for PI88941:
GlycoLink Columns, 1mL, 10 columns
GlycoLink Coupling Buffer, 2 x 100mL
Aniline, 0.2mL
Wash Buffer, 120mL
Sodium meta-periodate, 0.5g
Zeba Spin Desalting Columns, 5mL, 10 columns
PBS Pack (makes 500mL), 1 pack
Column Accessories, 10 ea
Kit Content for PI88942:
GlycoLink Columns, 0.1mL, 10 columns
GlycoLink Coupling Buffer, 100mL
Aniline, 0.2mL
Wash Buffer, 60mL
Sodium meta-periodate, 0.5g
Zeba Spin Desalting Columns, 0.5mL, 10 columns
PBS Pack (makes 500mL), 1 pack
Column Accessories, 10 ea
Kit Content is N/A for PI53149.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty