Would you like to log in again?
GlycoLink IP Kit, Thermo Scientific
Supplier: Thermo Scientific
88943 88943
PI88943EA 876.18 USD InStock
CAPI88943 PI88943
GlycoLink IP Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Immunoprecipitate using antibodies immobilized by their Fc-region glycosylation.
The Thermo Scientific GlycoLink IP Kit provides components for effective immunoprecipitation (IP and co-IP) based on small-scale, covalent, affinity-resin immobilization of antibodies and other glycoproteins via oxidized sugar groups.
The GlycoLink Kit uses hydrazide-activated UltraLink Resin to immobilize and perform 25 IP or Co-IP assays with different antibodies or glycoprotein ligands. The method is ideal for polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Monoclonal antibodies that contain adequate carbohydrates are also effective. Antibodies immobilized by this hydrazide-to-carbohydrate method have unobstructed antigen-binding sites and optimal purification capability. The prepared glycoprotein or antibody columns can be regenerated and reused at least five times for immunoprecipitation, co-immunoprecipitation or pull-down affinity-purification without significant loss in binding capacity.
Immunoprecipitation followed by electrophoresis and excision of the protein for mass spectrometry or sequencing. Immunoprecipitation and SDS-PAGE analysis of a protein with a molecular weight similar to the antibody’s heavy or light chain. Identification of binding partners of immobilized glycoprotein. Co-immunoprecipitation for determining protein-protein interactions. The GlycoLink IP Kit method results in covalent attachment of oxidized sugar groups of an IP antibody (or any other glycoprotein with oxidizable carbohydrate groups) to the hydrazide resin. The antibody resin is then incubated with the sample that contains the protein antigen of interest, allowing the antibody-antigen complex to form. After washing to remove non-bound components of the sample, the antigen is recovered by dissociation from the antibody with elution buffer supplied in the kit. The entire procedure is performed in a microcentrifuge spin cup, allowing solutions to be fully separated from the resin with a brief centrifugation. Only the antigen is eluted by the procedure, enabling it to be identified and further analyzed without interference from antibody fragments.
Ordering information: Kit contains resin, buffers and reagents for immobilization; desalting columns; IP lysis/wash and elution buffers for immunoprecipitation; microcentrifuge spin columns and collection tubes; and loading buffer for SDS-PAGE. Kit contains UltraLink Hydrazide Resin, 0.5mL; GlycoLink Coupling Buffer, 50mL; Aniline, 0.2mL; Sodium meta-periodate, 0.5g; Wash Buffer, 60mL; Zeba Spin Desalting Columns,0.5mL, 25 columns; IP Lysis/Wash Buffer, 50mL; PBS Pack (makes 500mL), 1 pack; Elution Buffer, pH 2.8, 50mL; Lane Marker Sample Buffer, Non-reducing, (5X), 5mL; Pierce Spin Columns-Screw Caps, 25 columns; Collection Tubes, 2mL, 100 tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty