Would you like to log in again?
Pierce™ Mitochondria Isolation Kit for Tissue, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
89801 89801
PI89801EA 466.52 USD InStock
CAPI89801 PI89801
Pierce™ Mitochondria Isolation Kit for Tissue, Thermo Scientific
Protein Extraction Protein Extraction Kits
The Mitochondria Isolation Kit for Tissue enables isolation of intact mitochondria from soft and hard tissue samples.
  • Quick and convenient - isolate intact mitochondria in less than 60 minutes
  • Versatile - offers two methods of isolation from both soft and hard tissue samples
  • Multi-sample format - reagent-based approach enables simultaneous processing of multiple samples
  • Optional alternate method - reagents and protocol included for the traditional Dounce homogenization procedure with fewer required strokes
  • Bench-top compatible - both procedures can be performed using a microcentrifuge tube
Both reagent based and Dounce homogenization procedures use differential centrifugation to separate the intact mitochondria using a bench top microcentrifuge and are completed in less than 60 minutes. In addition, both procedures have been optimized for maximum yield of mitochondria with minimal damage to integrity. The isolated mitochondria may be used for many downstream applications, including 1D and 2D western blotting and protein profiling using mass spectroscopy.
Delivery information: Kit contains Mitochondria isolation reagent A (50 mL), Mitochondria isolation reagent B (0.5 mL), Mitochondria isolation reagent C (65 ml), Bovine Serum Albumin (235 mg) and PBS Pack (makes 500 ml), 1 pack.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty