Would you like to log in again?
Pierce™ Protein Stabilizing Cocktail, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
89806 89806
PI89806EA 219.69 USD InStock
PI89806 CAPI89806
Pierce™ Protein Stabilizing Cocktail, Thermo Scientific
Protein Purification Protein Purification Reagents
A versatile stabilizing solution that increases the shelf-life of purified or partially purified proteins during routine storage.
  • Stabilizes most enzymes and other proteins significantly better than ordinary buffers
  • Protect proteins from environmental stresses
  • Does not destabilize biomolecules in downstream assays
  • Components are low molecular weight and fully dialyzable
  • Low-viscosity reagent is easier to pipette than 50 % glycerol
The formulation is comprised of low-weight, naturally occurring molecules that protect from environmental stresses that could lead to enzyme inactivation, aggregation, and freeze-thaw damage
Protein classes tested include kinases, phosphatases, peroxidases, restriction enzymes, luciferases, cytokines and antibodies.
In addition, it will not destabilize biomolecules in downstream assays. However, if needed, all components of the reagent can be removed by dialysis or desalting before the sample proteins are used in downstream assays. The solution is a 4X concentrate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty