Would you like to log in again?
Pierce™ Biotin 3' End DNA Labeling Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
89818 89818
PI89818EA 630.65 USD InStock
PI89818 CAPI89818
Pierce™ Biotin 3' End DNA Labeling Kit, Thermo Scientific
Protein Linking and Labeling Systems
Kit has been optimized to incorporate 1-3 biotinylated ribonucleotides (biotin-N4-CTP) onto the 3' end of DNA strands where the label will be less likely to interfere with hybridization or sequence-specific binding of proteins.
  • Non isotopic labeling eliminates the hassle of hazardous radioactive materials or difficult-to-dispose-of waste
  • 1-3 biotinylated ribonucleotides onto the 3' end of DNA strands for less interference with hybridisation or sequence-specific binding of proteins
  • Biotin-labeled probes are stable for more than one year
  • Fast, efficient 30-minute labeling procedure
The 30-minute procedure uses non-isotopic labeling components to eliminate the hassle of hazardous radioactive materials and the expense of special disposal costs. The resulting probes are stable for up to one year and can be used to facilitate non-isotopic detection in a variety of applications including electrophoretic mobility shift assays (EMSA)(see PI20148 series), northern or Southern blots, colony hybridizations, or in situ hybridizations. For fast and easy protein extraction kits, see PI89826 series.
Delivery information: The kit contains sufficient reagents for 20 reactions (each with 5 pmol DNA) and includes 5X TdT Reaction Buffer (1 mL); TdT (terminal deoxynucleotidyl tranferase), (5 µL); Biotin-11-UTP, (1 µL); Unlabeled control oligo, (14 µL); and Biotin-control oligo, (4 µL).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty