Would you like to log in again?
B-PER™ Pierce™ Complete Bacterial Protein Extraction Reagent, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce™
89822 89821 89821 89822
PI89821EA 488.06 USD InStock
CAPI89821 PI89821 CAPI89822 PI89822
B-PER™ Pierce™ Complete Bacterial Protein Extraction Reagent, Thermo Scientific
Protein Extraction Protein Extraction Reagents
B-PER™ Complete Bacterial Protein Extraction Reagent is an all-in-one formulation of non ionic detergent and enzymes that enables mild extraction of proteins from both gram-negative and gram-positive bacteria without the need for mechanical disruption. B-PER Complete Reagent combines a universal nuclease and lysozyme to improve cell lysis and reduce viscosity, and may be used for soluble protein extraction and inclusion body purification from bacterial cell lysates.
  • Fast and simple - just add B-PER Reagent to a bacterial pellet, incubate with mixing for 10 to 15 minutes and recover soluble proteins after pelleting the cell debris
  • Convenient - contains lysozyme and a universal nuclease in a single formulation with 4°C storage
  • Excellent yields - recover recombinant proteins from bacterial lysates or purify inclusion bodies to near-homogeneous levels
  • Suitable for any scale of protein extraction
  • Completely compatible with addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (eg. GST, 6xHis) and other applications
B-PER™ Complete Bacterial Protein Extraction Reagents are compatible with downstream applications, such as affinity chromatography (eg. immobilised metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification. Depending on the particular application, protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty