Would you like to log in again?
Pierce Pro-Ject™ Protein Transfection Reagent, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce™, Pro-Ject™
89850 89850
PI89850EA 666.83 USD InStock
CAPI89850 PI89850
Pierce Pro-Ject™ Protein Transfection Reagent, Thermo Scientific
Transfection Reagents
This Pierce™ protein transfection reagent is a cationic lipid mixture for complexation with proteins, peptides, antibodies and other biologically active molecules to allow their direct intracellular delivery.
  • No need to isolate, clone and transfect gene sequences
  • Optimum delivery in three to four hours after incubation
  • Delivery complex is non-covalent
  • Works with proteins, peptides and fluorescently labeled antibodies
The reagent/protein complexes offer optimum delivery within three to four hours after incubation and the complex is noncovalent so it does not interfere with biological activity
Molecules that have been successfully delivered with the transfection reagent include β-galactosidase, fluorescent-antibody, green fluorescent protein (GFP), high and low molecular weight dextran sulfate, phycoerythrin-BSA, caspase 3, caspase 8 and granzyme B.
It attaches to negatively charged cell surfaces and enters the cell either by directly fusing with the plasma membrane or by enodcytosis and subsequent fusion with the endosome. The transfection reagent may be used to study intercellular signaling, cell cycle regulation, apoptosis, oncogenesis, and transcriptional regulation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty