Would you like to log in again?
Pierce™ In-Gel Tryptic Digestion Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
89871 89871
PI89871EA 385.28 USD InStock
CAPI89871 PI89871
Pierce™ In-Gel Tryptic Digestion Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
The Pierce™ In-Gel Tryptic Digestion Kit provides a complete set of reagents for digesting and recovering peptide fragments from stained protein bands excised from SDS-PAGE (polyacrylamide gel plugs).
  • Contains highly purified and modified MS-grade trypsin that shows no chymotryptic activity and minimal autolytic activity
Includes all necessary reagents for destaining Coomassie®- or fluorescent dye-stained proteins, reduction and alkylation of cystines, and tryptic digestion. Procedure and reagents produce reliable digestions and data generation using a wide range of conditions without optimization.
Delivery information: The kit includes modified porcine trypsin, destaining buffers, reduction reagents, alkylation reagents and digestion buffers along with detailed and simple instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty