Would you like to log in again?
Pierce™ Nucleic Acid Detection Blocking Buffer, Thermo Scientific
Supplier: Thermo Scientific
Pierce
89880A
PI89880AEA 334.81 USD InStock
PI89880A
Pierce™ Nucleic Acid Detection Blocking Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Nucleic Acid Detection Blocking Buffer is intended for use with the Thermo Scientific Chemiluminescent Nucleic Acid Detection Module Kit, of which it is also a component
  • Fast and sensitive
  • Versatile kit protocol (included) is applicable to any biotin-based nucleic acid detection system: Northerns, Southerns, EMSA, RPAs and colony lifts
  • Kit contains only the critical detection components for those who already have biotin-labeled probes and an optimised hybridisation method developed
Non-specific binding of probe to membrane can be minimized by incubating the membrane in blocking buffer for 15 minutes. The system uses an enhanced luminol substrate for horseradish peroxidase (HRP) with optimized blocking and wash steps that together produce sensitivity equivalent to radioactive (32P) systems and provide a clean, consistent background required for publication-quality data.
It is a complete system for the detection of biotin-labeled nucleic acids for various blotting applications, including Northern/Southern blots, ribonuclease protection assays (RPAs), and electrophoretic mobility shift assays (EMSAs) or gel-shift assays. Store at 4˚C.
Delivery information: Kit contains stabilized Streptavidin-HRP conjugate, 1,5 mL; chemiluminescent substrate (luminol/enhancer solution, 80 mL and stable peroxide solution, 80 mL); blocking buffer, 500 mL; 4X wash buffer, 500mL; and substrate equilibration buffer, 500mL.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty