Would you like to log in again?
Pierce™ In-Solution Tryptic Digestion and Guanidination Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
89895 89895
PI89895EA 653.91 USD InStock
CAPI89895 PI89895
Pierce™ In-Solution Tryptic Digestion and Guanidination Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
Pierce™ In-Solution Tryptic Digestion and Guanidination Kit contains proteomics grade trypsin, buffers and reagents to alkylate and digest proteins and then guanidinate the peptide fragments for mass spectrometry.
  • Optimized trypsin digestion producing complete digests of protein ranging from 0.025 to 10 µg
  • Optimized guanidination where reaction produces little or no N-terminal modification
  • Protein can be digested and guanidinated all in a day
Accurate identification of proteins and analysis of post-translational modifications by mass spectrometry require accurate and complete protein digestion and peptide modification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty