Would you like to log in again?
Pierce™ Centrifuge Columns for Affinity Chromatography, Thermo Scientific
Supplier: Thermo Scientific
Pierce
89898 89896 89898 89897 89897 89868 89869 89869 89868 89896
PI89868EA 107.62 USD InStock
CAPI-89869 PI89868 CAPI89896 CAPI89898 CAPI89897 PI89897 PI-89869 PI89898 CAPI89868 PI89896
Pierce™ Centrifuge Columns for Affinity Chromatography, Thermo Scientific
Filters Centrifugal Filters
Pierce™ Centrifuge Columns are convenient tools for handling 40 µl to 10 ml of an affinity support for purification.
  • Made from low protein binding polypropylene for compatibility with protein purification
  • Filter type, pore size: Polyethylene filter, ~30 µm pore size
  • Twist-off bottom
These centrifuge columns can be used in either spin or gravity-flow format.
Applications:
Affinity purification, affinity chromatography
Immunodepletion
Ion exchange resins
Gel filtration, desalting, buffer exchange
The spin format reduces sample processing time and makes column washing and elution more efficient. For gravity-flow procedures and when slow processing time is desired for binding steps with dilute samples, Column Extenders can be used to increase the column reservoir capacity. Use the any combination of processing techniques to get the best possible chromatography results. Store at room temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty