Would you like to log in again?
Pierce™ Ubiquitin Enrichment Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
89899 89899
PI89899EA 850.34 USD InStock
CAPI89899 PI89899
Pierce™ Ubiquitin Enrichment Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
The Thermo Scientific Pierce Ubiquitin Enrichment Kit facilitates the isolation of polyubiquitin protein conjugates from cultured cells and tissue samples.
  • Fast—less than 45 minutes hands-on time for binding and recovery of ubiquitinated proteins
  • Complete—includes all reagents needed for ubiquitin-modified protein enrichment from cultured cells and tissue samples
  • Flexible—sample incubation from 2 hours to overnight allows assay to be completed in several hours or in less than 30 minutes after overnight incubation
  • Robust—compatible with all Thermo Scientific Pierce Cell Lysis Products and standard RIPA formulations
  • Multiple-sample format—easily processes 1 to 15 samples concurrently
The enriched fraction can subsequently be analyzed be quantitatively or qualitatively, depending on the application. The spin-column protocol requires less than 45 minutes of hands-on prep time and incubation can range from two hours to overnight for flexibility in scheduling assay times. The components in the kit are compatible with all Poppers cell lysis products and standard RIPA formulations. It will process up to 15 samples concurrently.
Delivery information: The kit contains Polyubiquitin Positive control (1000X, 50 µL), Anti-Ubiquitin Antibody (Rabbit antiserum, 5 µL), Polyubiquitin affinity resin slurry (300 µL), TBS pack (makes 500 mL), and Spin Columns (18) and Accessories.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty