Would you like to log in again?
n-Dodecyl-beta-Maltoside Detergent, Thermo Scientific
Supplier: Thermo Scientific
Pierce™
89902 89903 89902 89903
PI89902EA 218.4 USD InStock
PI89902 PI89903 CAPI89902 CAPI89903
n-Dodecyl-beta-Maltoside Detergent, Thermo Scientific
Cleaning Agents Detergents and Surface Cleansers
n-Dodecyl-β-D-Maltoside is especially useful for solubilizing membrane proteins to preserve their activity. Store below -20˚C.
  • Lipid-like nonionic detergent
  • Especially useful for isolating and stabilizing hydrophobic membrane proteins
  • Preserves activity of membrane protein better than most of the detergents
  • High-purity compound with low UV absorptivity
The water-soluble nonionic detergent is most often used for the isolation of hydrophobic membrane proteins
Like most detergents, n-Dodecyl-β-D-maltoside has dual hydrophobic/hydrophilic properties that facilitate lipid displacement and provide a lipid-like environment for membrane proteins. Studies suggest that the ability of these surfactants to preserve membrane protein structure stems in part from the reduced disruption of lipid:protein interactions where some of the natural lipid associations are maintained. While no detergent or set of conditions is optimal for all studies on membrane proteins, n-Dodecyl-β-D-maltoside has unique characteristics that have made it especially helpful in certain protein methods.
Multiple studies have shown that n-Dodecyl-β-D-maltoside is a gentle detergent that is often able to preserve protein activity better than many commonly used detergents, including NP-40, CHAPS and Octyl-β-glucoside.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty