Would you like to log in again?
Pierce™ Affinity Purification Kits, NAb™ Spin Kits and Columns for Antibody Purification, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
89963 89963 89979 89981 89958 89951 89978 89959 89980 89961 89948 89956 89956 89961 89950 89958 89952 89960 89959 89949 89978 89980 89953 89950 89953 89960 89957 89962 89957 89981 89949 89962 89979 89954 89954 89951 89955 89955 89952
PI89980EA 527.26 USD InStock
PI89980 PI89963 CAPI89955 CAPI89954 CAPI89979 CAPI89957 CAPI89978 CAPI89956 PI89981 CAPI89959 CAPI89958 PI89960 PI89961 PI89962 CAPI89980 CAPI89960 CAPI89981 PI89956 PI89978 CAPI89962 CAPI89961 PI89957 PI89979 PI89958 CAPI89963 PI89959 PI89952 PI89953 PI89954 PI89955 PI89950 PI89951 CAPI89949 PI89949 CAPI89951 CAPI89950 CAPI89953 PI89948 CAPI89952
Pierce™ Affinity Purification Kits, NAb™ Spin Kits and Columns for Antibody Purification, Thermo Scientific
Protein Purification Protein Purification Kits
The Pierce™ NAb™ Spin Kits are convenient for rapid, small-scale affinity purification of antibodies from a variety of sample types. Each pre-filled microcentrifuge spin column of the immobilized protein resin enables quick purification of 100 to 1000 µg of IgG from 25 to 500 µL of serum or other sample. The actual amount of IgG purified varies depending upon the sample type and the specific spin column used.
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior manufacturing method immobilizes by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
Proteins A and G bind to many of the same species and subclasses of IgG, although they have particular differences in affinity and binding capacity
Protein L binds to certain immunoglobulin kappa light chains. Because kappa light chains occur in members of all immunoglobulin classes (i.e., IgG, IgM, IgA, IgE and IgD), Protein L can purify these different classes of antibody. However, only those antibodies within each class that possess the appropriate kappa light chains will bind. Generally, empirical testing is required to determine if Protein L is effective for purifying a particular antibody.
Protein A is generally preferred for affinity purification of rabbit, pig, dog and cat IgG. Protein G has better binding capacity for a broader range of mouse and human IgG subclasses (IgG1, IgG2, etc.). Protein A/G is a recombinant fusion protein that includes the IgG-binding domains of both Protein A and Protein G. Therefore, Protein A/G is ideal for binding the broadest range of IgG subclasses from rabbit, mouse, human and other mammalian samples.
Delivery information: Each NAb™ Spin Kit, 0.2 mL contains spin columns (10), microcentrifuge tubes, 2 mL (80), Binding Buffer (50 mL), IgG Elution Buffer (1 pack, 500 mL) and Neutralisation Buffer (12 mL). Spin columns also available separately. Each NAb™ Spin Kit, 1 mL, contains spin columns (2), Binding Buffer (1 pack, 500 mL), IgG Elution Buffer (240 mL) and Neutralisation Buffer (12 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty