Would you like to log in again?
Pierce™ Phosphoprotein Enrichment Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
90003 90003
PI90003EA 956.31 USD InStock
CAPI90003 PI90003
Pierce™ Phosphoprotein Enrichment Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
Phosphoprotein Enrichment Kit is designed for the efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues.
  • Specific – low nonspecific protein contamination from complex biological samples, such as cell culture lysate and mouse tissue extract
  • Fast – easy spin format enables enrichment of phosphorylated proteins in less than 2 hours
  • Superior yield – acheives higher yields than other commercially available kits
  • Convenient format – complete kit offering pre-dispensed spin columns, buffers, reagents and ultrafiltration columns for enrichment
  • Compatible – downstream applications include mass spectrometry, Western blotting, and 2D-PAGE
The proprietary resin and buffer composition produces superior yields with negligible non-specific binding. The kit is uses a convenient spin column format and is compatible with downstream applications including Western blotting, Mass Spec, 2D-PAGE, and protein arrays. The affinity resin is also available as pre-packed chromatography cartridges for use with automated liquid chromatography (LC) systems.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty