Would you like to log in again?
Pierce™ Trypsin Protease, MS Grade, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
90057 90059 90057 90058 90058 90305 90305 90059
PI90305EA 88.92 USD InStock
CAPI90059 CAPI90058 PI90305 CAPI90057 CAPI90305 PI90059 PI90057 PI90058
Pierce™ Trypsin Protease, MS Grade, Thermo Scientific
Protein Extraction Protein Extraction Reagents
Pierce™ Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
  • Cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
  • No detectable chymotrypsin activity thus making it highly pure
  • Chemically modified for reduced autolytic activity
Trypsin is a serine protease derived from porcine pancreatic extracts. Applications include in-gel digestion of proteins from 1-D or 2-D gels and in-solution tryptic digestion of proteins. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. It is TPCK-treated to eliminate chymotryptic activity and methylated to improve stability during protein digestion. The enzyme is most active in pH ranges 7 to 9 and can be reversibly inactivated at pH <4.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty