Would you like to log in again?
Pierce™ iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
90103 90103
PI90103PK 3778.85 USD InStock
CAPI90103 PI90103
Pierce™ iodoTMTsixplex™ Isobaric Mass Tag Labeling Kit, Thermo Scientific
Protein Linking and Labeling Systems
Thermo Scientific iodoTMTsixplex Isobaric Mass Tag Labeling Kit contains iodoTMT™ reagents and associated materials for the irreversible labeling of sulfhydryl groups.
  • Specifically reacts with sulfhydryl groups (reduced, unmodified cysteine residues)
  • Labeled proteins and peptides are not susceptible to reducing agents
  • Options for duplex isotopic (MS) or sixplex isobaric (MS/MS) quantitation
  • Can be used to study cysteine modifications (e.g. di-sulfide bridges, S-nitrosylation)
Thermo Scientific iodoTMTsixplex Label Reagents enables enable concurrent identification and multiplexed quantitation of proteins in different samples using tandem mass spectrometry. The iodoTMT™ Reagents are sets of isobaric (mass and structure) isomers that are iodoacetyl-activated for covalent, irreversible labeling of sulfhydryl (—SH) groups. Similar to iodoacetamide, iodoTMT Reagents react specifically with reduced cysteines (Cys) in peptides and proteins. IodoTMT Reagents can be differentiated by mass spectrometry (MS), enabling quantitation of the relative abundance of cysteine modifications, such as S-nitrosylation, oxidation and disulfide bonds, in cultured cells grown or treated with different conditions.
Delivery information: Kit includes 5 x 0.2 mg iodoTMT6-126 Label Reagent,, iodoTMT6-127 Label Reagent, iodoTMT6-128 Label Reagent, iodoTMT6-129 Label Reagent, iodoTMT6-130 Label Reagent, iodoTMT6-131 Label Reagent, HES Buffer (15 mL), Bond-Breaker TCEP Solution (0.5 mL), DTT No-Weigh Format (8 x 7.7 mg), Trypsin (5 x 20 µg), Trypsin Storage Solution (250 µL), Bovine Albumin (2.5 mg).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty