Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydroxylamine solution 50% (w/w) in water for TMT experiments

Supplier: 0000013509
Danger

Pierce
90115 90115
PI90115EA 126.77 USD InStock
CAPI90115 PI90115
Hydroxylamine solution 50% (w/w) in water for TMT experiments
Hydroxylamine
Thermo Scientific 50% Hydroxylamine is a 10X quenching reagent for amine-reactive isobaric mass tag labeling experiments.

Used to prepare oximes, and is also an intermediate in biological nitrification.

Caution: For research use only. Not for use in diagnostic procedures.


Formula: H₃NO
MW: 33.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044522
CAS Number: 7803-49-8
EINECS: 232-259-2
UN: 3266
ADR: 8,III
Order Now
Specification Test Results
ASSAY/TITRATION 45.0 - 55.0% range
VISUAL Colorless/clear to slightly yellow liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty