Would you like to log in again?
Pierce™ MS-Compatible Magnetic IP Kit, protein A/G, Thermo Scientific
Supplier: Thermo Fisher Scientific
90409
PI90409EA 736 USD InStock
PI90409
Pierce™ MS-Compatible Magnetic IP Kit, protein A/G, Thermo Scientific
Protein Linking and Labeling Systems
The Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kit (Protein A/G) provides mass spectrometry-friendly reagents and an optimized protocol to enable highly effective and efficient immunoprecipitation and co-immunoprecipitation of target antigens upstream of LC-MS analysis.MS-compatible—reagents directly compatible with in-solution peptide digestion, enriched samples contain minimal detergent residuals detected using LC-MSSensitive—procedure successfully enriches low abundance proteins (low ng range)Low background—binding, wash, and elution buffers optimized to minimize enrichment of background proteinsRobust—procedure and reagents have been robustly tested with numerous targets to ensure consistent enrichment of low abundance proteins (ng range) with at least 2 peptides identified per proteinThe Pierce MS-Compatible Magnetic IP Kit (Protein A/G) uses high-quality Pierce Protein A/G Magnetic Beads to provide wider flexibility of antibody capture than either Protein A or G alone
The optimized kit components have been formulated to be compatible with downstream LC-MS analysis. After the immunoprecipitation procedure, the target-enriched elution fraction is ready for in-solution tryptic digestion without the need for gel purification, detergent removal, or desalting.This kit has been rigorously validated using numerous target antigens with varying expression levels, including targets previously undetected without enrichment or by western blotting. Additionally, the reagents and procedures have been validated using both manual and automated magnetic separation.Applications:Enrichment of low abundance targets to improve MS sensitivityIdentification of target proteins that are not detectable by western blottingQuantitation of target proteins by LC-MS analysisScreening of antibodies for IP-MSEnrichment of target proteins upstream of targeted analysis (SRM)
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Protein A/G Magnetic Beads, 1 mL
IP-MS Cell Lysis Buffer, 100 mL
IP-MS Wash Buffer A, 75 mL
IP-MS Wash Buffer B, 40 mL
IP-MS Elution Buffer, 6 mL
MgCl2 (1 M), 0.75 mL
Store in refrigerator (2–8˚C).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty