Would you like to log in again?
Pierce 3UV Lamp, Thermo Scientific
Supplier: Thermo Scientific
Pierce
95035 95034 95034 95035
PI95034EA 1095.88 USD InStock
CAPI95035 CAPI95034 PI95034 PI95035
Pierce 3UV Lamp, Thermo Scientific
Lights UV Lights, Lamps and Bulbs
Thermo Scientific Pierce UV Lamps are compact, multifunctional ultraviolet lamps with 252 nm, 302 nm and 365 nm dial-selectable wavelength settings especially for use with photoactivatable crosslinkers and UV-crosslinking methods.
  • Unique reflector design provides maximum UV for all applications
  • Designed for hand-held or stationary use
  • Lamp features aluminum construction and scratch-resistant finish
  • 8-watt lamps are ideal for a wide area of illumination, such as culture plates
  • This lamp is validated for use in crosslinking applications with diazirine and phenyl azide photoactivatable crosslinkers
15.5 L x 3 W x 4.5" D (394 x 76 x 114 mm).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty