Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Streptavidin, HRP (Horseradish Peroxidase), Pierce Chemical

Supplier: Pierce Chemical
N100 N100
PIN100EA 176.51 USD InStock
PIN100 CAPIN100
Streptavidin, HRP (Horseradish Peroxidase), Pierce Chemical
Proteins and Peptides
HRP-Conjugated Streptavidin consists of streptavidin protein that is covalently conjugated to horseradish peroxidase (HRP) enzyme (RZ > 3.0). Streptavidin binds to biotin and the conjugated HRP provides enzyme activity for detection using an appropriate substrate system. This particular product has been used primarily in sandwich ELISA applications to provide consistent measurement of biotinylated detection antibodies.

Formulation: Provided in a conjugate stabilizer containing 0.1% ProClin Compound as a preservative.
Shipped on cold packs.
Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is from 1:4,000 to 1:20,000. The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.

Ordering information: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty