Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Streptavidin, Poly-HRP (Horseradish Peroxidase), Pierce Chemical

Supplier: Pierce Chemical
N200 N200
PIN200EA 341.17 USD InStock
PIN200 CAPIN200
Streptavidin, Poly-HRP (Horseradish Peroxidase), Pierce Chemical
Proteins and Peptides
Poly-HRP Streptavidin consists of streptavidin protein that is covalently conjugated to a polymerized form of horseradish peroxidase (HRP) enzyme. Streptavidin binds to biotin and the conjugated HRP polymer provides a high level of activity for detection using an appropriate substrate system. This particular Poly-HRP Streptavidin product has been used primarily in sandwich ELISA applications to improve the signal provided by typical (non-Poly) streptavidin-HRP conjugates.

Features of Poly-HRP Streptavidin:

Formulation: Conjugate in PBS containing distilled glycerol, BSA and a preservative.
Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is 1:2,000 - 1:20,000. The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.

Formulation: Conjugate in PBS containing distilled glycerol, BSA and a preservative.
Application: ELISA: to be used with biotin-labeled detection antibodies. Recommended dilution range is 1:2,000 - 1:20,000. The optimal dilution must be determined empirically for each specific application. Prepare dilutions immediately before use.

Ordering information: For Research Use Only. Not for use in diagnostic procedures. Do not store below -20˚C. Avoid multiple freeze-thaw cycles.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty