Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TMB Substrate Solution, Thermo Scientific

Supplier: Thermo Scientific
N301 N301
PIN301EA 158.74 USD InStock
PIN301 CAPIN301
TMB Substrate Solution, Thermo Scientific
Assays ELISAs
Addition of sulfuric acid stop solution changes the color to yellow, enabling accurate measurement of the intensity at 450nm using a spectrophotometer or plate reader. This particular formulation is used in most of the Thermo Scientific Pierce ELISA Kits where it provides consistent performance for the sandwich ELISA technique.

Features of TMB Substrate Solution:

Product Number: N301
Size: 100 mL (sufficient for about ten 96-well microplates)
Storage: Shipped on cold packs. Store at 2-8˚C immediately upon receipt. Do not use product beyond the stated expiration date.
Application: ELISA: Recommended for peroxidase-based enzyme immunoassays. Upon oxidation, TMB forms a water-soluble blue reaction product that can be measured spectrophotometrically at 605nm. Upon acidification, the reaction product becomes yellow with an absorbance peak at 450nm.
Precautions: This product is sensitive to contamination from a variety of oxidizing agents. To avoid contamination and premature expiration, avoid contacting TMB solution with any potential source of contamination. Never pipette directly from the bottle. Pour out required amount onto a tube and pipette from the tube. Do not return excess TMB to primary storage container.

Procedure:
Add 100 µL of TMB substrate to each well containing peroxidase reaction.
Incubate at room temperature for 15 to 30 minutes.
Add 100 µL of stop solution (0.16M sulfuric acid,

Product Number: N301

Ordering information: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty