Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Poly HRP Dilution Buffer, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

N501 N501
PIN501EA 1010.4 USD InStock
PIN501 CAPIN501
Poly HRP Dilution Buffer, Thermo Scientific
Buffers Assay Buffers
Thermo Scientific Pierce Poly-HRP Dilution Buffer is optimized to ensure optimal performance with Poly-HRP Streptavidin.

Features of Poly-HRP Dilution Buffer:

Formulation: 1% biotin-free casein in a PBS buffer solution.
Application: Dilution of Poly-HRP Streptavidin (Product # N200): Gently invert bottle and swirl to mix before use. Do not vortex. Use at full strength. Refer to Poly-HRP Streptavidin Certificate of Analysis for recommended dilution ranges. Dilute Poly-HRP Streptavidin immediately before use.

Formulation: 1% biotin-free casein in a PBS buffer solution.

For Research Use Only. Not for use in diagnostic procedures.

Ordering information: Shipped on cold packs. Store at 2-80˚C immediately upon receipt.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty