Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stop Solution for TMB Substrates, Thermo Scientific

Supplier: Thermo Scientific
N600 N600
PIN600EA 128.83 USD InStock
PIN600 CAPIN600
Stop Solution for TMB Substrates, Thermo Scientific
Assays ELISAs
Stop Solution is 0.16M sulfuric acid for use with the ELISA substrate 3,3’,5,5’ - tetramethylbenzidine (TMB). In the presence of horseradish peroxidase (HRP) enzyme conjugates, TMB and peroxide react to produce a blue byproduct having maximum absorbance at 605nm. The color intensity is proportional to the amount of HRP activity, which in turn is related to the levels of target analyte in an optimized ELISA procedure. Addition of sulfuric acid stop solution changes the color to yellow (absorbance maximum at 450nm), stabilizing the color development to enable accurate measurement of the intensity at 450nm using a spectrophotometer or plate reader.

Features of Stop Solution:

Size: 55 mL (sufficient for about five 96-well microplates)
Storage: Shipped at ambient temperature. Store at room temperature or 2-8˚C.
Application: ELISA: Following TMB incubation, add an amount of stop solution equal to the amount of TMB substrate added to each well of the ELISA plate (typically 50 - 100 mL per well). Use at full strength.

Size: 55 mL (sufficient for about five 96-well microplates)

Ordering information: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty