Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Premium Grade, Pierce™

Supplier: 0000013509
Warning
Synonyms: EDAC hydrochlorideEDAC HClEDC HClEDC1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochlorideWSC HCl1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochlorideN-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochlorideEDC hydrochlorideN'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride

PG82074 PG82079 PG82074 PG82079 PG82073 PG82073
PIPG82079EA 134.91 USD InStock
CAPIPG82073 CAPIPG82074 PIPG82079 CAPIPG82079 PIPG82074 PIPG82073
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Premium Grade, Pierce™
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Thermo Scientific Pierce Premium Grade EDC is our highest quality formulation of this popular carbodiimide crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.

  • EDC—carbodiimide crosslinker for conjugation between carboxylates and amines
  • High quality—identity and purity confirmed by several tests
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review

Compared to the standard grade product, Premium Grade EDC provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. EDC is 1-ethyl-3-[3-dimethylaminopropyl]carbodiimide hydrochloride, a water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation. In the presence of N-hydroxysulfosuccinimide (sulfo-NHS), EDC can be used to convert carboxyl groups to amine-reactive sulfo-NHS esters. This is accomplished by mixing the EDC with a carboxyl containing molecule and adding sulfo-NHS.

Specifications of Premium Grade EDC
We manufacture crosslinking reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.
• Visual: White crystalline powder, free of foreign matter
• Identity: IR scan shows only peaks characteristic of EDC
• Water content: Karl Fischer titration, ≤2%
• Trace metals: All metals, including lead, ≤50 ppm
• Elemental analysis: Reported values for C, H, and N

Caution: For Research Use Only. Not for use in diagnostic procedures.


Formula: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/32/20961432.jpg
Order Now
Specification Test Results
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short
Form Solid
Quantity 1 g/25 g/500 g
Type Crosslinker
Solubility Water

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty