Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium pyroantimonate solution R USP, Reag. Ph. Eur. 1071301

Supplier: Ricca Chemical
Danger

R6459000-500A
R6459000500AEA 195.75 USD InStock
R6459000500A
Potassium pyroantimonate solution R USP, Reag. Ph. Eur. 1071301
Potassium pyroantimonate solution R
Potassium Pyroantimonate Solution R EP Reagent 1071301 / USP for Saccharin Sodium
Suitability for Use: dense precipitate with Sodium
Potassium Pyroantimonate Solution R ref: EP
Potassium Pyroantimonate Solution ref: USP


Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0200 g/cm³
Storage Temperature: Ambient
UN: 3266
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear to hazy, colorless, odorless
Suitability for Use dense precipitate with Sodium

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty