Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromophenol blue 0.1% (w/v) in isopropanol (IPA) indicator

Supplier: Ricca Chemical
Synonyms: Bromphenol Blue Sultone FormTetrabromophenol-sulfonephthalein3,3',5,5'-TetrabromophenolsulfonphthaleinTetrabromophenol-sulphonephthaleinBPB3,3',5,5'-Tetrabromophenolsulphonphthalein

RSOD9800-500A
RC-SOD98-16EA 397.73 USD InStock
RC-SOD98-16
Bromophenol blue 0.1% (w/v) in isopropanol (IPA) indicator
Bromophenol blue


Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
MDL Number: MFCD00005875
CAS Number: 115-39-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty