Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Biuret reagent solution USP Test Solution (TS)

Supplier: Ricca Chemical
Warning

R1025000-1A R1025000-500A
RC1025-16EA 99.67 USD InStock
RC1025-16 RC1025-32
Biuret reagent solution USP Test Solution (TS)
Biuret reagent solution


Storage Temperature: Ambient
UN: 3266
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, blue, odorless
Suitability for Use Violet color in presence of Pepsin

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty