Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brij® 35 30% (w/w) aqueous wetting agent

Supplier: Ricca Chemical
Danger
Synonyms: Brij® L23Tricosaethylene glycol mono-n-dodecyl etherPolyoxyethylene(23)lauryl ether

R1145000-1A
RC1145-32EA 97.94 USD InStock
RC1145-32
Brij® 35 30% (w/w) aqueous wetting agent
Brij® 35


Formula: C₅₈H₁₁₈O₂₄
MW: 1199.56 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0300 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00080891
CAS Number: 9002-92-0
Order Now
Specification Test Results
Appearance Clear, colorless, soapy solution with characteristic odor
Refractive Index Confirmed concentration

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty