Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brilliant cresyl blue 1% (w/v) in 0.85% sodium chloride stain for reticulocyte count

Supplier: Ricca Chemical
Synonyms: BCBBrilliant Blue CBrilliant cresyl blue zinc chloride double salt1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1)CI 51010

R1150000-500A R1150000-120A
RC11504EA 58.23 USD InStock
RC11504 RC115016
Brilliant cresyl blue 1% (w/v) in 0.85% sodium chloride stain for reticulocyte count
Brilliant Cresyl Blue


Formula: C₃₄H₄₂Cl₄N₈O₂Zn
MW: 801.96 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00148901
CAS Number: 81029-05-2
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Bluish-purple, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty