Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Brilliant green 1% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning
Synonyms: CI 42040Basic Green 1N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulfateBrilliant Green hydrogen sulfateBrilliant Green hydrogen sulphateDiamond GreenAstradiamant green GXN-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphateSolid Green JOEthyl GreenMalachite Green GEmerald Green

R1155000-500A R1155000-120A
RC1155-4EA 59.12 USD InStock
RC1155-4 RC1155-16
Brilliant green 1% (w/v) in aqueous solution
Brilliant Green


Formula: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011880
CAS Number: 633-03-4
Order Now
Specification Test Results
Appearance Clear, dark teal, odorless
Suitability for Use Yellow (pH 2.6)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty