Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromate-Bromide solution 0.0250 N (N/40)

Supplier: Ricca Chemical
Warning
Synonyms: Bromide-Bromate

R1166000-4C R1166000-1C
RC1166-32EA 124.33 USD InStock
RC1166-32 RC1166-1
Bromate-Bromide solution 0.0250 N (N/40)
Bromate-Bromide


Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.02500 ± 0.00002 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty