Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromine number titration solvent dichloromethane formulation

Supplier: Ricca Chemical
Danger

R1191000-2.5D R1191000-4C1
RC1191-1EA 640.29 USD InStock
RC1191-1 RC1191-5PT
Bromine number titration solvent dichloromethane formulation
Bromine number titration solvent


Density: 1.0590 g/cm³
Storage Temperature: Ambient
UN: 2790
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, Acetic Acid odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty