Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromine water (saturated solution of bromine in water) 3% (w/v) ACS

Supplier: Ricca Chemical

R1195000-1C1 R1195000-500C1
RC119532EA 532.84 USD InStock
RC119532 RC119516
Bromine water (saturated solution of bromine in water) 3% (w/v) ACS
Bromine water (saturated solution of bromine in water, ca. 9%)


Formula: Br₂
MW: 159.81 g/mol
MDL Number: MFCD00010896
CAS Number: 7726-95-6
UN: 1744
ADR: 8,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty