Would you like to log in again?
Bromocresol green-methyl red in aqueous solution mixed indicator
Supplier: Ricca Chemical
Synonyms: Methyl Red - Bromocresol green solution
R1215000-500A R1215000-120A R1215000-1A
RC1215-16EA 96.93 USD InStock
RC1215-16 RC1215-4 RC1215-32
Bromocresol green-methyl red in aqueous solution mixed indicator
Bromocresol green-methyl red
Order Now
Specification Test Results
Appearance Dark green-blue, odorless
Suitability for Use Greenish-Blue (pH 5.4) - Light Pink (pH 4.5), with Gray warning color
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty