Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol green-methyl red solution alcoholic solution mixed indicator

Supplier: Ricca Chemical
Danger
Synonyms: Methyl Red - Bromocresol green solution

R1220000-1A R1220000-120A R1220000-500A
RC122016EA 138.9 USD InStock
RC122016 RC1220-32 RC12204
Bromocresol green-methyl red solution alcoholic solution mixed indicator
Bromocresol green-methyl red


Boiling Pt: ~ 93 °C
Density: 0.9000 g/cm³
Flash Pt: ~ 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00134126
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Dark green, alcohol odor
Suitability for Use Greenish-Blue (pH 5.4) - Light Pink (pH 4.5), with Gray warning color

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty