Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol green-methyl red USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: Methyl Red - Bromocresol green solution

R1221000-500A R1221000-120A
RC1221-16EA 135.03 USD InStock
RC1221-16 RC1221-4
Bromocresol green-methyl red USP Test Solution (TS)
Bromocresol green-methyl red


Boiling Pt: ~ 77 °C
Density: 0.8300 g/cm³
Flash Pt: ~ 69°F
Storage Temperature: Ambient
MDL Number: MFCD00134126
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, red, alcohol odor
Suitability for Use Greenish-Blue (pH 5.2) - Light Pink (pH 4.5), with Gray warning color

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty