Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol purple 0.04% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Synonyms: 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide5',5''-Dibromo-o-cresolsulphonphthalein5',5''-Dibromo-o-cresolsulfonphthalein

R1300000-500A R1300000-120A
RC130016EA 59.59 USD InStock
RC130016 RC13004
Bromocresol purple 0.04% (w/v) in aqueous solution indicator
Bromocresol purple


Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Order Now
Specification Test Results
Appearance Red, odorless
Suitability for Use Yellow (pH 6.8)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty