Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromophenol blue 0.1% (w/v) in dilute alcohol USP Test Solution (TS)

Supplier: Ricca Chemical
Danger
Synonyms: Bromphenol Blue Sultone FormTetrabromophenol-sulfonephthalein3,3',5,5'-TetrabromophenolsulfonphthaleinTetrabromophenol-sulphonephthaleinBPB3,3',5,5'-Tetrabromophenolsulphonphthalein

R1357000-500A R1357000-120A
RC1357-16EA 230.37 USD InStock
RC1357-16 RC1357-4
Bromophenol blue 0.1% (w/v) in dilute alcohol USP Test Solution (TS)
Bromophenol blue


Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
UN: 1170
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, reddish color, ethanol odor
Suitability for Use Yellow (pH 4.6)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty